missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropionitrile fumarate, 99+%

Product Code. 10182890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10182890 25 g Plastic bottle 25g
10353522 5 g Glass bottle 5g
10529620 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10182890 Supplier Thermo Scientific Acros Supplier No. 104470250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2079-89-2 | 0·5 C4H4O4 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2079-89-2
Molecular Formula 0·5 C4H4O4
Molecular Weight (g/mol) 128.13
MDL Number MFCD00078321

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 170.0°C to 173.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula H2NCH2CH2CN·1/2HO2CCH=CHCO2H
Quantity 25 g
Formula Weight 128.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopropionitrile fumarate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminopropionitrile fumarate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.