missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropionitrile fumarate, 98+%

Product Code. 11417126
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417126 25 g 25g
11407126 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417126 Supplier Thermo Scientific Alfa Aesar Supplier No. B25440.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2079-89-2 | (NH2CH2CH2CN)2·HO2CCH=CHCO2H

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2079-89-2
Molecular Formula (NH2CH2CH2CN)2·HO2CCH=CHCO2H
MDL Number MFCD00078321
Synonym Bis(3-aminopropionitrile) fumarate; 2-Cyanoethylamine hemifumarate

Specifications

Melting Point ∼177°C (decomposition)
Odor Odorless
Quantity 25 g
Beilstein 3573532
Formula Weight 256.26
Percent Purity ≥98%
Chemical Name or Material 3-Aminopropionitrile fumarate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.