missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropionitrile fumarate (2:1), MedChemExpress

Product Code. 16415126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16415126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16415126 Supplier Medchem Express Supplier No. HY107829

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-Aminopropionitrile fumarate (2:1) is a lathyrogen which inhibits crosslinking of collagen.

3-Aminopropionitrile fumarate (2:1) is a lathyrogen which inhibits crosslinking of collagen.

Specifications

Chemical Name or Material 3-Aminopropionitrile fumarate (2:1)
CAS 2079-89-2
Color White
Purity Grade Notes Research
Molecular Formula C4H4O4.2C3H6N2
Quantity 100 mg
Synonym Di-β-aminopropionitrile fumarate β-Aminopropionitrile fumarate β-Ammoniumpropionitrile hemifumarate
Solubility Information H2O : 125 mg/mL (487.79 mM; Need ultrasonic)
SMILES NCCC#N.OC(/C=C/C(O)=O)=O.NCCC#N
Molecular Weight (g/mol) 256.26
Formula Weight 256.26
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.