missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropionitrile, 98%, stab. with potassium carbonate

Product Code. 11479973
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479973 1 g 1g
11489973 5 g 5g
11499973 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479973 Supplier Thermo Scientific Alfa Aesar Supplier No. A13043.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 151-18-8 | C3H6N2 | 70.10 g/mol

3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.

Solubility
Soluble in water.

Notes
It is air and moisture sensitive. Incompatible with oxidizing agents, acids and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-18-8
Molecular Formula C3H6N2
Molecular Weight (g/mol) 70.10
MDL Number MFCD00014820
InChI Key AGSPXMVUFBBBMO-UHFFFAOYSA-N
Synonym 3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino
PubChem CID 1647
ChEBI CHEBI:27413
IUPAC Name 3-aminopropanenitrile
SMILES NCCC#N

Specifications

Density 0.952
Boiling Point 79°C to 81°C (16 mmHg)
Odor Amine-like
Linear Formula H2NCH2CH2CN
Refractive Index 1.4375
Quantity 1 g
UN Number UN2735
Beilstein 1698848
Sensitivity Air and moisture sensitive
Merck Index 14,469
Solubility Information Soluble in water.
Formula Weight 70.1
Percent Purity 98%
Chemical Name or Material 3-Aminopropionitrile, stabilized with potassium carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.