missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopicolinic Acid, TRC

Product Code. 30314770
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314770 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314770 Supplier Toronto Research Chemicals Supplier No. TRCA6278005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Aminopicolinic Acid
CAS 1462-86-8
Molecular Formula C6 H6 N2 O2
Quantity 5 g
Synonym 2-Pyridinecarboxylic acid, 3-amino-, Picolinic acid, 3-amino- (6CI,7CI,8CI), 3-Amino-2-pyridinecarboxylic acid, 3-Amino-2-carboxypyridine, 3-Amino-2-picolinic acid, 3-Aminopicolinic acid
InChI Formula InChI=1S/C6H6N2O2/c7-4-2-1-3-8-5(4)6(9)10/h1-3H,7H2,(H,9,10)
SMILES Nc1cccnc1C(=O)O
IUPAC Name 3-aminopyridine-2-carboxylic acid
Molecular Weight (g/mol) 138.12
Formula Weight 138.0429
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.