missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophthalic acid hydrochloride dihydrate, 96%

Product Code. 11464083
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464083 5 g 5g
11454083 1 g 1g
11474083 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464083 Supplier Thermo Scientific Alfa Aesar Supplier No. A10319.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1852533-96-0 | C8H8ClNO4 | 217.605 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1852533-96-0
Molecular Formula C8H8ClNO4
Molecular Weight (g/mol) 217.605
MDL Number MFCD00150282
InChI Key ZBZAVEORKXFUQB-UHFFFAOYSA-N
Synonym 3-aminophthalic acid hydrochloride, 1,2-benzenedicarboxylic acid, 3-amino-, hydrochloride, 1,2-benzenedicarboxylic acid, 3-amino-, hydrochloride 1:1, phthalic acid, hydrochloride, 1, 3-amino-, hydrochloride, 3-aminobenzene-1,2-dicarboxylic acid hydrochloride, 3-amino phthalic acid hydrochloride, phthalic acid, 3-amino-, hydrochloride, acmc-1b7k3, ghl.pd_mitscher_leg0.1315
PubChem CID 81375
IUPAC Name 3-aminophthalic acid;hydrochloride
SMILES C1=CC(=C(C(=C1)N)C(=O)O)C(=O)O.Cl

Specifications

Melting Point 182°C (decomposition)
Quantity 5 g
Formula Weight 253.65 (217.62 Anhydrous)
Percent Purity 98%
Chemical Name or Material 3-Aminophthalic acid hydrochloride dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.