missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenylacetylene, 98%

Product Code. 11374566
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374566 5 g 5g
11384566 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374566 Supplier Thermo Scientific Alfa Aesar Supplier No. L20124.06
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 54060-30-9 | C8H7N | 117.15 g/mol

It is used as Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as Pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heavy metal powders.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54060-30-9
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00014779
InChI Key NNKQLUVBPJEUOR-UHFFFAOYSA-N
Synonym 3-aminophenylacetylene, benzenamine, 3-ethynyl, m-aminophenylacetylene, 3-ethynylbenzenamine, 3-ethynylphenylamine, 3-ethynylaniline, m-aminophenyl acetylene, 3-aminophenyl acetylene, m-ethynylaniline, 3-ethynyl-aniline
PubChem CID 104682
IUPAC Name 3-ethynylaniline
SMILES NC1=CC=CC(=C1)C#C

Specifications

Melting Point 22°C
Density 1.04
Boiling Point 70°C (1 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.617
Quantity 5 g
Beilstein 2935417
Solubility Information Insoluble in water.
Formula Weight 117.15
Percent Purity 98%
Chemical Name or Material 3-Aminophenylacetylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.