missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophenol, 98+%

Product Code. 11413944
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413944 5000 g 5000g
11403944 250 g 250g
11493934 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413944 Supplier Thermo Scientific Alfa Aesar Supplier No. A11943.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-27-5 | C6H7NO | 109.13 g/mol

A chemical used in the preparation of inhibitors of mammalian carbonic anhydrase isoforms. It is also used in the preparation of quinoline tethered fluorescent carbon dots for regulated anticancer discovery. It is employed in the production of TB drugs, salicylic acid, amino stabilizer and developers. 3-Aminophenol has been used as a stabilizer of chlorine-containing thermoplastics, although its principal use is as an intermediate in the production of 4-amino-2-hydroxybenzoic acid this isomer is also employed as a hair colorant and as a coupler molecule in hair dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A chemical used in the preparation of inhibitors of mammalian carbonic anhydrase isoforms. It is also used in the preparation of quinoline tethered fluorescent carbon dots for regulated anticancer discovery. It is employed in the production of TB drugs, salicylic acid, amino stabilizer and developers. 3-Aminophenol has been used as a stabilizer of chlorine-containing thermoplastics, although its principal use is as an intermediate in the production of 4-amino-2-hydroxybenzoic acid this isomer is also employed as a hair colorant and as a coupler molecule in hair dyes.

Solubility
Soluble in water (35 mg/ml at 20°C), ether, alcohol, amyl alcohol, and benzene (slightly).

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-27-5
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00007786
InChI Key CWLKGDAVCFYWJK-UHFFFAOYSA-N
Synonym m-aminophenol, m-hydroxyaniline, 3-hydroxyaniline, phenol, 3-amino, 1-amino-3-hydroxybenzene, 3-amino-1-hydroxybenzene, fouramine eg, futramine eg, fourrine eg, pelagol eg
PubChem CID 11568
ChEBI CHEBI:28924
IUPAC Name 3-aminophenol
SMILES NC1=CC=CC(O)=C1

Specifications

Melting Point 122°C to 126°C
Density 0.99
pH 6.8
Boiling Point 164°C (11 mmHg)
Flash Point 178°C (352°F)
Odor Characteristic
Quantity 5000 g
UN Number UN2512
Beilstein 636059
Merck Index 14,460
Solubility Information Soluble in water (35mg/ml at 20°C),ether,alcohol,amyl alcohol,and benzene (slightly).
Formula Weight 109.13
Percent Purity ≥98%
Chemical Name or Material 3-Aminophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.