missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopentane, 98%

Product Code. 15488799
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488799 5 g 5g
11391557 25 g 25g
11301567 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15488799 Supplier Thermo Scientific Alfa Aesar Supplier No. L02250.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 616-24-0 | C5H13N | 87.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-24-0
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.17
MDL Number MFCD00008096
InChI Key PQPFFKCJENSZKL-UHFFFAOYSA-N
Synonym 3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane
PubChem CID 12019
ChEBI CHEBI:84248
IUPAC Name pentan-3-amine
SMILES CCC(N)CC

Specifications

Density 0.748
Boiling Point 89°C to 91°C
Flash Point 2°C (35°F)
Odor Amine-like
Refractive Index 1.4055
Quantity 5 g
UN Number UN1106
Beilstein 635658
Sensitivity Air Sensitive
Formula Weight 87.17
Percent Purity 98%
Chemical Name or Material 3-Aminopentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.