missing translation for 'onlineSavingsMsg'
Learn More

3-Aminomethyl-benzoic acid methyl ester hydrochloride, 95%, Thermo Scientific™

Product Code. 10462644
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10462644 10 g Amber Glass Bottle 10g
10300593 250 mg Amber Glass Bottle 250mg
10433113 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10462644 Supplier Thermo Scientific Maybridge Supplier No. MO07352EA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 17841-68-8
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
InChI Key UOWRPTFJISFGPI-UHFFFAOYSA-N
Synonym methyl 3-aminomethyl benzoate hydrochloride, 3-aminomethylbenzoic acid methyl ester hydrochloride, 3-aminomethyl-benzoic acid methyl ester hydrochloride, methyl 3-aminomethylbenzoate hcl, methyl 3-amino methyl benzoate hydrochloride, methyl 3-aminomethylbenzoate hydrochloride, methyl 3-aminomethyl benzoate hcl, benzoic acid, 3-aminomethyl-, methyl ester, hydrochloride, methyl 3-aminomethyl benzoate, chloride, pubchem11178
PubChem CID 2794825
IUPAC Name methyl 3-(aminomethyl)benzoate;hydrochloride
SMILES COC(=O)C1=CC(=CC=C1)CN.Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 201.65
Percent Purity 95%
Chemical Name or Material 3-Aminomethylbenzoic acid methyl ester hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.