missing translation for 'onlineSavingsMsg'
Learn More

3-(Aminomethyl)benzoic acid hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15431819
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 876-03-9
Molecular Formula C8H10ClNO2
Molecular Weight (g/mol) 187.623
MDL Number MFCD03791117
InChI Key SJCCOASSOPUHEN-UHFFFAOYSA-N
Synonym 3-aminomethyl benzoic acid hydrochloride, 3-aminomethylbenzoic acid hydrochloride, 3-aminomethylbenzoic acid hcl, 3-aminomethyl-benzoic acid hydrochloride, acmc-209tzp, 3-aminomethyl benzoic acid hcl, 3-carboxybenzylamine hydrochloride, 3-aminomethyl-benzoic acid hcl salt, 3-aminomethyl benzoic acid, chloride, 3-aminomethyl benzoicacid hydrochloride
PubChem CID 16218772
IUPAC Name 3-(aminomethyl)benzoic acid;hydrochloride
SMILES C1=CC(=CC(=C1)CN)C(=O)O.Cl

Specifications

Melting Point 242°C to 244°C
Quantity 1 g
Beilstein 3697656
Solubility Information Slightly soluble in water.
Formula Weight 187.62
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(Aminomethyl)benzoic acid hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'