missing translation for 'onlineSavingsMsg'
Learn More

3-(Aminomethyl)benzeneboronic acid hydrochloride, 96%

Product Code. 15490546
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490546 250 mg 250mg
15480546 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490546 Supplier Thermo Scientific Alfa Aesar Supplier No. H53284.MD
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 352525-94-1 | C7H11BClNO2 | 187.43 g/mol

3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352525-94-1
Molecular Formula C7H11BClNO2
Molecular Weight (g/mol) 187.43
MDL Number MFCD02093050
InChI Key NPTBTFRGCBFYPZ-UHFFFAOYSA-N
Synonym 3-aminomethyl phenyl boronic acid hydrochloride, 3-aminomethyl phenylboronic acid hydrochloride, 3-aminomethylphenylboronic acid hydrochloride, 3-aminomethylphenyl boronic acid hydrochloride, 3-aminomethylphenylboronic acid hcl, 3-aminomethylphenyl boronic acid hcl, boronic acid, 3-aminomethyl phenyl-, hydrochloride, 3-aminomethylphenylboronic acid, hcl
PubChem CID 16427085
IUPAC Name [3-(aminomethyl)phenyl]boronic acid;hydrochloride
SMILES B(C1=CC(=CC=C1)CN)(O)O.Cl

Specifications

Melting Point 248°C to 250°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 187.43
Percent Purity 96%
Chemical Name or Material 3-(Aminomethyl)benzeneboronic acid hydrochloride
compliance-icons
Product Identifier
  • 3-(Aminomethyl)benzeneboronic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 3-(Aminomethyl)benzeneboronic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.