missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoisonicotinic acid, 98%, Thermo Scientific™

Product Code. 15525266 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15525266 5 g 5g
15515266 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15525266 Supplier Thermo Scientific Acros Supplier No. 457690050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7579-20-6
Molecular Formula C6H5N2O2
Molecular Weight (g/mol) 137.12
MDL Number MFCD00137840
InChI Key FYEQKMAVRYRMBL-UHFFFAOYSA-M
Synonym 3-aminoisonicotinic acid, 3-amino-4-pyridinecarboxylic acid, 3-amino-isonicotinic acid, 3-amino-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 3-amino, 3-amino-4-carboxypyridine, 3-amino-4-pyridinecarboxylicacid, 3-amino-4-carboxylic acid pyridine, pubchem2420, 3-aminoisonicotinsaure
PubChem CID 459531
IUPAC Name 3-aminopyridine-4-carboxylic acid
SMILES NC1=CN=CC=C1C([O-])=O

Specifications

Melting Point 275.0°C to 276.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 3-Aminoisonicotinic acid
compliance-icons
Product Identifier
  • 3-Aminoisonicotinic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.