missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocoumarin, 97%

Product Code. 11358905
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
Unit Size:
100mg
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358905 500 mg 500mg
11368905 100 mg 100mg
11499830 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358905 Supplier Thermo Scientific Alfa Aesar Supplier No. L14031.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1635-31-0 | C9H7NO2 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1635-31-0
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00016965
InChI Key QWZHDKGQKYEBKK-UHFFFAOYSA-N
Synonym 3-aminocoumarin, 3-amino-2h-chromen-2-one, 3-amino-2-benzopyrone, 3-amino-2h-1-benzopyran-2-one, coumarin amine, coumarin monoamine, amino-chromen-2-one, 3-azanylchromen-2-one, acmc-1bous
PubChem CID 74217
IUPAC Name 3-aminochromen-2-one
SMILES C1=CC=C2C(=C1)C=C(C(=O)O2)N

Specifications

Melting Point 135°C to 137°C
Quantity 500 mg
Beilstein 128033
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 3-Aminocoumarin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.