missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocoumarin, 97%, Thermo Scientific Chemicals

Product Code. 11368905
Click to view available options
Quantity:
100 mg
500 mg
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1635-31-0
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00016965
InChI Key QWZHDKGQKYEBKK-UHFFFAOYSA-N
Synonym 3-aminocoumarin, 3-amino-2h-chromen-2-one, 3-amino-2-benzopyrone, 3-amino-2h-1-benzopyran-2-one, coumarin amine, coumarin monoamine, amino-chromen-2-one, 3-azanylchromen-2-one, acmc-1bous
PubChem CID 74217
IUPAC Name 3-aminochromen-2-one
SMILES C1=CC=C2C(=C1)C=C(C(=O)O2)N

Specifications

Melting Point 135°C to 137°C
Quantity 100 mg
Beilstein 128033
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 3-Aminocoumarin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'