missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzyl alcohol, 97%

Product Code. 10387600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10387600 5 g Glass bottle 5g
10390254 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10387600 Supplier Thermo Scientific Acros Supplier No. 184570050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1877-77-6 | C7H9NO | 123.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1877-77-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00007817
InChI Key OJZQOQNSUZLSMV-UHFFFAOYSA-N
Synonym 3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol
PubChem CID 80293
IUPAC Name (3-aminophenyl)methanol
SMILES NC1=CC=CC(CO)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 91.0°C to 95.0°C
Color Beige or Beige-Gray to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CH2OH
Quantity 5 g
Beilstein 13, 619
Formula Weight 123.15
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-Aminobenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.