missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminobenzotrifluoride, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 3-Trifluoromethylaniline |
| CAS | 98-16-8 |
| Molecular Formula | C7 H6 F3 N |
| Quantity | 100 g |
| Synonym | 3-(Trifluoromethyl)aniline, Niflumic Acid Imp. C (EP), Flutamide Imp. D (EP), Benzenamine, 3-(trifluoromethyl)- (9CI, ACI), 3-(Trifluoromethyl)benzenamine (ACI), m-Toluidine, α,α,α-trifluoro- (6CI, 7CI, 8CI), 1-Amino-3-(trifluoromethyl)benzene, 3-(Trifluoromethyl)phenylamine, 3-Amino-α,α,α-trifluorotoluene, 3-Aminobenzotrifluoride, m-(Trifluoromethyl)aniline, m-Amino-α,α,α-trifluorotoluene, m-Aminobenzal fluoride, m-Aminobenzotrifluoride, m-Trifluoromethylaniline, NSC 4540, α,α,α-Trifluoro-m-toluidine |
| InChI Formula | InChI=1S/C7H6F3N/c8-7(9,10)5-2-1-3-6(11)4-5/h1-4H,11H2 |
| SMILES | Nc1cccc(c1)C(F)(F)F |
| IUPAC Name | 3-(trifluoromethyl)aniline |
| Molecular Weight (g/mol) | 161.12 |
| Formula Weight | 161.0452 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction