missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzotrifluoride, 98%

Product Code. 10090540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10090540 500 mL Glass bottle 500mL
10725361 5 mL Glass bottle 5mL
10667242 100 mL Glass bottle 100mL
10035923 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10090540 Supplier Thermo Scientific Acros Supplier No. 103195000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-16-8 | C7H6F3N | 161.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-16-8
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.13
MDL Number MFCD00007797
InChI Key VIUDTWATMPPKEL-UHFFFAOYSA-N
Synonym 3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline
PubChem CID 7375
IUPAC Name 3-(trifluoromethyl)aniline
SMILES C1=CC(=CC(=C1)N)C(F)(F)F

Specifications

CAS Min % 98
Melting Point 5.0°C to 6.0°C
Color Yellow
Density 1.3030g/mL
Boiling Point 187.0°C to 193.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CF3
Refractive Index 1.4790 to 1.4810
Quantity 500 mL
Beilstein 12, 870
Specific Gravity 1.303
Solubility Information Solubility in water: 5g/L in water (20°C). Other solubilities: soluble in ethanol,ether and diethyl ether
Formula Weight 161.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Aminobenzotrifluoride, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Aminobenzotrifluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.