missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzophenone, 98%, Thermo Scientific™

Product Code. 10268703 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10268703 1 g Glass bottle 1g
10066153 5 g Glass bottle 5g
10197083 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10268703 Supplier Thermo Scientific Acros Supplier No. 392860010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2835-78-1
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00017103
InChI Key FUADXEJBHCKVBN-UHFFFAOYSA-N
Synonym 3-aminobenzophenone, 3-benzoylaniline, methanone, 3-aminophenyl phenyl, 3-aminophenyl phenyl methanone, benzophenone, 3-amino, m-aminobenzophenone, 3-aminophenyl phenylmethanone, 3-aminophenyl phenyl ketone, 3-amino-benzophenon, 3-amino-benzophenone
PubChem CID 17817
IUPAC Name (3-aminophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 84.0°C to 87.0°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(O)C6H4NH2
Quantity 1 g
Formula Weight 197.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Aminobenzophenone
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminobenzophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.