missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzoic Acid, TRC

Product Code. 30136444
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136444 100 g 100g
30011794 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136444 Supplier Toronto Research Chemicals Supplier No. TRCA591490100G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-05-8
Molecular Formula C7 H7 N O2
Molecular Weight (g/mol) 137.14
Synonym 3-Aminobenzoic Acid, Mesalazine Imp. D (EP), Benzoic acid, 3-amino- (9CI, ACI), 3-Aminobenzoic acid (ACI), Benzoic acid, m-amino- (8CI), 3-Aminobenzenecarboxylic acid, 3-Carboxyaniline, Aniline-3-carboxylic acid, m-Aminobenzoic acid, m-Anthranilic acid, m-Carboxyaniline, m-Carboxyphenylamine, MeSH ID: D062368, NSC 15012
IUPAC Name 3-aminobenzoic acid
SMILES Nc1cccc(c1)C(=O)O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C7H7NO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,8H2,(H,9,10)
Formula Weight 137.0477
Chemical Name or Material 3-Aminobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.