missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzeneboronic acid, 98%

Product Code. 15419687
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15419687

Brand: Thermo Scientific Alfa Aesar H52909.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30418-59-8 | C6H8BNO2 | 136.945 g/mol

3-Aminobenzeneboronic acid is used as an organic chemical synthesis intermediate. It is a boronic acid with potential use for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzeneboronic acid is used as an organic chemical synthesis intermediate. It is a boronic acid with potential use for biochemical research.

Solubility
Soluble in water (partly miscible).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30418-59-8
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.945
MDL Number MFCD00007755
InChI Key JMZFEHDNIAQMNB-UHFFFAOYSA-N
Synonym 3-aminobenzeneboronic acid, 3-aminophenyl boronic acid, m-aminophenylboronic acid, 3-aminophenyl boranediol, m-aminophenyl boronic acid, 3-amino phenylboronic acid, chembl20852, boronic acid, 3-aminophenyl, m-aminophenyl metaboric acid
PubChem CID 92269
IUPAC Name (3-aminophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)N)(O)O

Specifications

Melting Point 138°C to 143°C
Quantity 5 g
Solubility Information Soluble in water (partly miscible).
Formula Weight 136.95
Percent Purity 98%
Chemical Name or Material 3-Aminobenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.