missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzene-1-sulfonamide, 97%, Thermo Scientific™

Product Code. 10160652
Change view
Click to view available options
Quantity:
1 g
25 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10160652 50 g Amber Glass Bottle 50g
10784013 1 g Amber Glass Bottle 1g
10123424 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10160652 Supplier Thermo Scientific Maybridge Supplier No. SB01949EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 98-18-0
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
InChI Key JPVKCHIPRSQDKL-UHFFFAOYSA-N
Synonym metanilamide, m-aminobenzenesulfonamide, benzenesulfonamide, 3-amino, m-sulfamoylaniline, 3-aminobenzene-1-sulfonamide, benzenesulfonamide, m-amino, m-aminobenzenesulphonamide, metaniilamide, 3-aminosulfonylaniline, 3-amino-benzenesulfonamide
PubChem CID 7377
IUPAC Name 3-aminobenzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula NH2C6H4SO2NH2
Quantity 50 g
Formula Weight 172.21
Percent Purity 97%
Chemical Name or Material 3-Aminobenzene-1-sulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.