missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzamide, MedChemExpress

Product Code. 16441626
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441626 500 mg 500mg
16441026 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16441626 Supplier Medchem Express Supplier No. HY12022
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 3-Aminobenzamide (PARP-IN-1) is a potent inhibitor of PARP with IC50 of appr 50 nM in CHO cells, and acts as a mediator of oxidant-induced myocyte dysfunction during reperfusion.

3-Aminobenzamide (PARP-IN-1) is a potent inhibitor of PARP with IC50 of appr 50 nM in CHO cells, and acts as a mediator of oxidant-induced myocyte dysfunction during reperfusion.

Specifications

Chemical Name or Material 3-Aminobenzamide
Physical Form Powder
CAS 3544-24-9
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C7H8N2O
For Use With (Application) Cancer-programmed cell death
Synonym PARP-IN-1
Solubility Information H2O : ≥ 11.11 mg/mL (81.60 mM)
SMILES O=C(C1=CC=CC(N)=C1)N
Molecular Weight (g/mol) 136.15
Formula Weight 136.15
Percent Purity 99.16%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.