missing translation for 'onlineSavingsMsg'
Learn More

3'-Aminoacetophenone, 97%

Product Code. p-7020519
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11474144

Brand: Thermo Scientific Alfa Aesar A12503.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-03-6 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-03-6
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00007796
InChI Key CKQHAYFOPRIUOM-UHFFFAOYSA-N
Synonym 3'-aminoacetophenone, 1-3-aminophenyl ethanone, m-aminoacetophenone, 3-aminoacetophenone, 3-acetylaniline, m-acetylaniline, ethanone, 1-3-aminophenyl, 1-3-aminophenyl ethan-1-one, m-aminoacetylbenzene, acetophenone, 3'-amino
PubChem CID 7417
IUPAC Name 1-(3-aminophenyl)ethanone
SMILES CC(=O)C1=CC=CC(N)=C1

Specifications

Melting Point 94°C to 98°C
Boiling Point 289°C to 291°C
Quantity 50 g
Beilstein 386009
Merck Index 14,413
Formula Weight 135.17
Percent Purity 97%
Chemical Name or Material 3'-Aminoacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.