missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-o-cresol, 95%, Thermo Scientific™

Product Code. 10032640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10032640 250 mg Glass bottle 250mg
10012630 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10032640 Supplier Thermo Scientific Acros Supplier No. 209042500
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 53222-92-7
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007787
InChI Key FLROJJGKUKLCAE-UHFFFAOYSA-N
Synonym 3-amino-o-cresol, phenol, 3-amino-2-methyl, 2-amino-o-cresol, 2-methyl-3-aminophenol, 3-hydroxy-2-methylaniline, aminokresol, o-cresol, 3-amino, pubchem15039, 2-methyl-3-amino-phenol, 3-amino-2-methyl phenol
PubChem CID 104448
IUPAC Name 3-amino-2-methylphenol
SMILES CC1=C(C=CC=C1O)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 127°C to 133°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H3(CH3)OH
Quantity 250 mg
Beilstein 13, 579
Formula Weight 123.15
Percent Purity 95%
Chemical Name or Material 3-Amino-o-cresol, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-o-cresol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.