Learn More
3-Amino-9-ethylcarbazole, 90%, tech., Thermo Scientific Chemicals
Chemical Identifiers
CAS | 132-32-1 |
---|---|
Molecular Formula | C14H14N2 |
Molecular Weight (g/mol) | 210.28 |
MDL Number | MFCD00004964 |
InChI Key | OXEUETBFKVCRNP-UHFFFAOYSA-N |
Synonym | 3-amino-9-ethylcarbazole, 9-ethyl-9h-carbazol-3-amine, 3-amino-n-ethylcarbazole, 9h-carbazol-3-amine, 9-ethyl, carbazole, 3-amino-9-ethyl, 9-ethylcarbazol-3-ylamine, unii-8q2bg27jbu, 9-ethylcarbazole-3-ylamine, 8q2bg27jbu, 9-ethyl-9h-carbazol-3-ylamine |
PubChem CID | 8588 |
IUPAC Name | 9-ethylcarbazol-3-amine |
SMILES | CCN1C2=C(C=C(C=C2)N)C3=CC=CC=C31 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10716351
|
Thermo Scientific Acros
147870050 |
5 g | Glass bottle |
N/A
|
N/A | ||||
10486970
|
Thermo Scientific Acros
147870250 |
25 g | Glass bottle |
N/A
|
N/A | ||||
10781745
|
Thermo Scientific Acros
147871000 |
100 g | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
132-32-1 | |
210.28 | |
OXEUETBFKVCRNP-UHFFFAOYSA-N | |
8588 | |
CCN1C2=C(C=C(C=C2)N)C3=CC=CC=C31 |
C14H14N2 | |
MFCD00004964 | |
3-amino-9-ethylcarbazole, 9-ethyl-9h-carbazol-3-amine, 3-amino-n-ethylcarbazole, 9h-carbazol-3-amine, 9-ethyl, carbazole, 3-amino-9-ethyl, 9-ethylcarbazol-3-ylamine, unii-8q2bg27jbu, 9-ethylcarbazole-3-ylamine, 8q2bg27jbu, 9-ethyl-9h-carbazol-3-ylamine | |
9-ethylcarbazol-3-amine |
Specifications
132-32-1 | |
100.0 | |
88% min. (GC) | |
MFCD00004964 | |
3-amino-9-ethylcarbazole, 9-ethyl-9h-carbazol-3-amine, 3-amino-n-ethylcarbazole, 9h-carbazol-3-amine, 9-ethyl, carbazole, 3-amino-9-ethyl, 9-ethylcarbazol-3-ylamine, unii-8q2bg27jbu, 9-ethylcarbazole-3-ylamine, 8q2bg27jbu, 9-ethyl-9h-carbazol-3-ylamine | |
OXEUETBFKVCRNP-UHFFFAOYSA-N | |
9-ethylcarbazol-3-amine | |
8588 | |
90% | |
3-Amino-9-ethylcarbazole |
88.0 | |
98.0°C to 100.0°C | |
C14H14N2 | |
22, IV, 4936 | |
Solubility in water: insoluble. | |
CCN1C2=C(C=C(C=C2)N)C3=CC=CC=C31 | |
210.28 | |
210.28 | |
Technical |
Safety and Handling
GHS H Statement
Toxic if swallowed.
May cause cancer.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
Wear protective gloves/protective clothing/e
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-057-7
missing translation for 'rtecsNumber' : FE3590000
missing translation for 'tsca' : TSCA
- 3-Amino-9-ethylcarbazole
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only