missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-tert-butyl-1H-pyrazole, 98%

Product Code. 11440723
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
missing translation for 'unitSize':
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
11440723 25 g 25g
11420723 1 g 1g
11430723 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11440723 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A12750.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82560-12-1 | C7H13N3 | 139.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82560-12-1
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.20
MDL Number MFCD00067987,MFCD00082631
InChI Key ZHBXGHWSVIBUCQ-UHFFFAOYSA-N
Synonym 3-amino-5-tert-butylpyrazole, 3-tert-butyl-1h-pyrazol-5-amine, 5-amino-3-tert-butylpyrazole, 3-amino-5-tert-butyl-1h-pyrazole, 5-tert-butyl-2h-pyrazol-3-ylamine, 5-amino-3-t-butylpyrazole, 1h-pyrazol-3-amine, 5-1,1-dimethylethyl, 5-tert-butyl-2h-pyrazol-3-amine, 5-amino-3-tert-butyl-1h-pyrazole
PubChem CID 522787
IUPAC Name 5-tert-butyl-1H-pyrazol-3-amine
SMILES CC(C)(C)C1=CC(N)=NN1

Specifications

Melting Point 75°C to 79°C
Quantity 25 g
Formula Weight 139.2
Percent Purity 98%
Chemical Name or Material 3-Amino-5-tert-butyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.