missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-tert-butyl-1H-pyrazole, 97%, ACROS Organics™

Product Code. 13585020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13585020 50 g Glass bottle 50g
13575020 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13585020 Supplier Thermo Scientific Acros Supplier No. 448650500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 82560-12-1
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.20
MDL Number MFCD00067987,MFCD00082631
InChI Key ZHBXGHWSVIBUCQ-UHFFFAOYSA-N
Synonym 3-amino-5-tert-butylpyrazole, 3-tert-butyl-1h-pyrazol-5-amine, 5-amino-3-tert-butylpyrazole, 3-amino-5-tert-butyl-1h-pyrazole, 5-tert-butyl-2h-pyrazol-3-ylamine, 5-amino-3-t-butylpyrazole, 1h-pyrazol-3-amine, 5-1,1-dimethylethyl, 5-tert-butyl-2h-pyrazol-3-amine, 5-amino-3-tert-butyl-1h-pyrazole
PubChem CID 522787
IUPAC Name 5-tert-butyl-1H-pyrazol-3-amine
SMILES CC(C)(C)C1=CC(N)=NN1

Specifications

Assay Percent Range 97%
Packaging Glass bottle
Quantity 50 g
Formula Weight 139.2
Chemical Name or Material 3-Amino-5-tert-butyl-1H-pyrazole
compliance-icons
Product Identifier
  • 3-Amino-5-tert-butyl-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.