missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-nitrobenzoic acid, 98%

Product Code. p-7020523
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11454154

Brand: Thermo Scientific Alfa Aesar A12508.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 618-84-8 | C7H6N2O4 | 182.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-84-8
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00017025
InChI Key ZNVHAQRPXAQKRU-UHFFFAOYSA-N
Synonym 3-amino-5-nitrobenzoic acid, 3-amino-5-nitro-benzoic acid, benzoic acid, 3-amino-5-nitro, 3-amino-5-nitrobenzoicacid, 5-amino-3-nitrobenzoic acid, acmc-20ana2, 3-nitro-5-aminobenzoic acid, 5-nitro-3-aminobenzoic acid, benzoic acid,3-amino-5-nitro, 3-amino-5-nitrobenzoic acid 1g
PubChem CID 239342
SMILES C1=C(C=C(C=C1N)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 214°C to 218°C
Quantity 1 g
Beilstein 1457935
Sensitivity Hygroscopic
Formula Weight 182.14
Percent Purity 98%
Chemical Name or Material 3-Amino-5-nitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.