missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-methyl-1H-pyrazole, 97%

Product Code. 11424753
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424753 25 g 25g
11414753 5 g 5g
11339519 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424753 Supplier Thermo Scientific Alfa Aesar Supplier No. A11642.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31230-17-8 | C4H7N3 | 97.121 g/mol

3-Amino-5-methyl-1H-pyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides. It is also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones. It can react with 4-Ethoxymethylene-2-phenyl-4H-oxazol-5-one to get N-(2-Methyl-7-oxo-4,7-dihydro-pyrazolo[1,5-a]pyrimidin-6-yl)-benzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-5-methyl-1H-pyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides. It is also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones. It can react with 4-Ethoxymethylene-2-phenyl-4H-oxazol-5-one to get N-(2-Methyl-7-oxo-4,7-dihydro-pyrazolo[1,5-a]pyrimidin-6-yl)-benzamide.

Solubility
Soluble in Dichloromethane and Methanol.

Notes
Air Sensitive. Store at -20°C. Store under dry inert gas away from oxidizing agents and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31230-17-8
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.121
MDL Number MFCD00075180
InChI Key FYTLHYRDGXRYEY-UHFFFAOYSA-N
Synonym 3-amino-5-methylpyrazole, 3-methyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-methyl, 5-amino-3-methylpyrazole, 3-amino-5-methyl-1h-pyrazole, 5-amino-3-methyl-1h-pyrazole, 5-methyl-2h-pyrazol-3-ylamine, 5-methyl-2h-pyrazol-3-amine, 5-methyl-1h-pyrazol-3-ylamine, 5-methylpyrazol-3-amine
PubChem CID 93146
IUPAC Name 5-methyl-1H-pyrazol-3-amine
SMILES CC1=CC(=NN1)N

Specifications

Melting Point 42°C to 47°C
Boiling Point 212°C to 213°C (14 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1904
Sensitivity Air Sensitive
Solubility Information Soluble in Dichloromethane and Methanol.
Formula Weight 97.12
Percent Purity 97%
Chemical Name or Material 3-Amino-5-methyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.