missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-hydroxybenzoic acid hydrochloride, 97%, Thermo Scientific™

Product Code. p-3540966
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10146433

Brand: Thermo Scientific Maybridge MO07975DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 14206-69-0
Molecular Formula C7H8ClNO3
Molecular Weight (g/mol) 189.60
MDL Number MFCD00043420
InChI Key CXESTILCPSBCGQ-UHFFFAOYSA-N
Synonym 3-amino-5-hydroxybenzoic acid hydrochloride, ahba hydrochloride, 3-amino-5-hydroxybenzoic acid hcl, 3-amino-5-hydroxy-benzoic acid hydrochloride, benzoic acid, 3-amino-5-hydroxy-, hydrochloride, 3-amino-5-carboxyphenol hydrochloride, 3-carboxy-5-hydroxyanilne hydrochloride, 3-amino-5-hydroxybenzoic acid, chloride, 3-amino-5-hydroxybenzoicacidhydrochloride, 3-amino-5-hydroxybenzoic acid-hydrogen chloride 1/1
PubChem CID 24229779
IUPAC Name 3-amino-5-hydroxybenzoic acid;hydrochloride
SMILES Cl.NC1=CC(O)=CC(=C1)C(O)=O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 207.62
Percent Purity 95%
Chemical Name or Material 3-amino-5-hydroxybenzoic acid hydrochloride hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.