missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-ethyl-1H-pyrazole, 97%

Product Code. 15295998 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15295998 1 g 1g
15206008 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15295998 Supplier Thermo Scientific Acros Supplier No. 446710010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1904-24-1 | C5H9N3 | 111.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1904-24-1
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD08060982
InChI Key AXDGPQLEVYSXNL-UHFFFAOYSA-N
Synonym 3-ethyl-1h-pyrazol-5-amine, 5-amino-3-ethyl-1h-pyrazole, 3-amino-5-ethylpyrazole, 3-amino-5-ethyl-1h-pyrazole, 5-ethyl-2h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 5-ethyl, 5-ethyl-2h-pyrazol-3-amine, ethyl-5-aminopyrazole, 3-amino-5-ethypyrazole, 1h-pyrazol-5-amine, 3-ethyl
PubChem CID 21991095
IUPAC Name 5-ethyl-1H-pyrazol-3-amine
SMILES CCC1=CC(N)=NN1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material 3-Amino-5-ethyl-1H-pyrazole
compliance-icons
Product Identifier
  • 3-Amino-5-ethyl-1H-pyrazole, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.