missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromopyridine, 98%, Thermo Scientific Chemicals

Catalog Number 15423068
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
13535-01-8
C5H5BrN2
173.013
MFCD01646060
MDQXGHBCDCOOSM-UHFFFAOYSA-N
3-amino-5-bromopyridine, 5-bromo-3-pyridinamine, 5-bromo-pyridin-3-ylamine, 5-bromo-3-aminopyridine, 3-pyridinamine, 5-bromo, 5-brompyridin-3-amin, 5-bromo-3-pyridylamine, 5-bromopyridin-3-ylamine, 3-amino-5-bromo pyridine, pubchem1213
817681
5-bromopyridin-3-amine
C1=C(C=NC=C1Br)N
This item is not returnable. View return policy
13535-01-8
C5H5BrN2
173.013
MFCD01646060
MDQXGHBCDCOOSM-UHFFFAOYSA-N
3-amino-5-bromopyridine, 5-bromo-3-pyridinamine, 5-bromo-pyridin-3-ylamine, 5-bromo-3-aminopyridine, 3-pyridinamine, 5-bromo, 5-brompyridin-3-amin, 5-bromo-3-pyridylamine, 5-bromopyridin-3-ylamine, 3-amino-5-bromo pyridine, pubchem1213
817681
5-bromopyridin-3-amine
C1=C(C=NC=C1Br)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 65°C to 69°C
Quantity 1 g
Formula Weight 173.01
Percent Purity 98%
Chemical Name or Material 3-Amino-5-bromopyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'