missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromo-2-hydroxypyridine, 95%, Thermo Scientific Chemicals

Product Code. 15458028
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98786-86-8
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.01
MDL Number MFCD03840438
InChI Key ULOVLVWNSQNABA-UHFFFAOYSA-N
Synonym 3-amino-5-bromopyridin-2-ol, 3-amino-5-bromo-2-hydroxypyridine, 2-hydroxy-3-amino-5-bromopyridine, 3-amino-5-bromopyridin-2 1h-one, 3-amino-5-bromo-2-pyridinol, 3-amino-5-bromo-pyridin-2-ol, 2 1h-pyridinone, 3-amino-5-bromo, pubchem6513, pubchem18742, acmc-20a0ks
PubChem CID 13434515
IUPAC Name 3-amino-5-bromo-1H-pyridin-2-one
SMILES NC1=CC(Br)=CNC1=O

Specifications

Quantity 1 g
Formula Weight 189.01
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Amino-5-bromo-2-hydroxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'