missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-(3,4-dimethoxyphenyl)-1,2,4-thiadiazole, 96%, Thermo Scientific™

Product Code. 15498755
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498755 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15498755 Supplier Thermo Scientific Alfa Aesar Supplier No. H34052.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1086385-71-8
Melting Point 159°C to 163°C
Molecular Formula C10H11N3O2S
MDL Number MFCD11655557
Quantity 500 mg
Synonym 5-3,4-dimethoxyphenyl-1,2,4-thiadiazol-3-amine, 3-amino-5-3,4-dimethoxyphenyl-1,2,4-thiadiazole
InChI Key WDEDMMNPBVPGJS-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)C2=NC(=NS2)N)OC
IUPAC Name 5-(3,4-dimethoxyphenyl)-1,2,4-thiadiazol-3-amine
Molecular Weight (g/mol) 237.277
PubChem CID 46741378
Formula Weight 237.28
Percent Purity 96%
Chemical Name or Material 3-Amino-5-(3,4-dimethoxyphenyl)-1,2,4-thiadiazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.