missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-(2-furyl)-1H-pyrazole, 97%

Product Code. 11372587
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372587 250 mg 250mg
11362587 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372587 Supplier Thermo Scientific Alfa Aesar Supplier No. L11721.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 96799-02-9 | C7H7N3O | 149.153 g/mol

3-Amino-5-(2-furyl)-1H-pyrazole is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-5-(2-furyl)-1H-pyrazole is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96799-02-9
Molecular Formula C7H7N3O
Molecular Weight (g/mol) 149.153
MDL Number MFCD00082696
InChI Key XJNZHICOWTVWOX-UHFFFAOYSA-N
Synonym 3-furan-2-yl-1h-pyrazol-5-amine, 3-2-furyl-1h-pyrazol-5-amine, 5-furan-2-yl-1h-pyrazol-3-amine, 5-amino-3-2-furyl pyrazole, 3-amino-5-2-furyl pyrazole, 5-2-furyl-1h-pyrazol-3-amine, 3-amino-5-2-furyl-1h-pyrazole, 5-2-furanyl-1h-pyrazol-3-amine, 5-amino-3-fur-2-yl-1h-pyrazole, 5-furan-2-yl-2h-pyrazol-3-amine
PubChem CID 523183
IUPAC Name 5-(furan-2-yl)-1H-pyrazol-3-amine
SMILES C1=COC(=C1)C2=CC(=NN2)N

Specifications

Melting Point 124°C to 128°C
Quantity 250 mg
Beilstein 4399373
Solubility Information Insoluble in water.
Formula Weight 149.15
Percent Purity 97%
Chemical Name or Material 3-Amino-5-(2-furyl)-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.