missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid, ≥95%, Thermo Scientific™

Product Code. 10163672
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10163672 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10163672 Supplier Thermo Scientific Maybridge Supplier No. SEW03132DA
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 849066-52-0
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C12H8F3NO2S
MDL Number MFCD00115029
Quantity 1 g
Synonym 3-amino-4-phenyl-5-trifluoromethyl thiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 3-amino-4-phenyl-5-trifluoromethyl
InChI Key BPFFXVCOJJGNQU-UHFFFAOYSA-N
SMILES NC1=C(SC(=C1C1=CC=CC=C1)C(F)(F)F)C(O)=O
IUPAC Name 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
Molecular Weight (g/mol) 287.26
PubChem CID 2740235
Formula Weight 287.26
Percent Purity 95%
Chemical Name or Material 3-Amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.