missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylpyridine, 98%

Product Code. 11395575
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395575 25 g 25g
11385575 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395575 Supplier Thermo Scientific Alfa Aesar Supplier No. L19730.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3430-27-1 | C6H8N2 | 108.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3430-27-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00128871
InChI Key IBKMZYWDWWIWEL-UHFFFAOYSA-N
Synonym 3-amino-4-methylpyridine, 3-amino-4-picoline, 3-pyridinamine, 4-methyl, 4-methyl-3-pyridinamine, 4-methyl-pyridin-3-ylamine, unii-9hn55rq07b, 4-methyl-3-pyridylamine, 4-methylpyridine-3-amine, 3-amino-4-methyl pyridine, 3-amino-4-methylpyridine 3-amino-4-picoline
PubChem CID 137935
IUPAC Name 4-methylpyridin-3-amine
SMILES CC1=C(C=NC=C1)N

Specifications

Melting Point 102°C to 106°C
Boiling Point 254°C
Odor Unpleasant
Quantity 25 g
UN Number UN2811
Beilstein 107792
Sensitivity Hygroscopic
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material 3-Amino-4-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.