missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylpyridine, 97%

Product Code. 10460768 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10460768 25 g Glass bottle 25g
10196337 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10460768 Supplier Thermo Scientific Acros Supplier No. 429860250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3430-27-1 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3430-27-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00128871
InChI Key IBKMZYWDWWIWEL-UHFFFAOYSA-N
Synonym 3-amino-4-methylpyridine, 3-amino-4-picoline, 3-pyridinamine, 4-methyl, 4-methyl-3-pyridinamine, 4-methyl-pyridin-3-ylamine, unii-9hn55rq07b, 4-methyl-3-pyridylamine, 4-methylpyridine-3-amine, 3-amino-4-methyl pyridine, 3-amino-4-methylpyridine 3-amino-4-picoline
PubChem CID 137935
IUPAC Name 4-methylpyridin-3-amine
SMILES CC1=C(C=NC=C1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 101.0°C to 102.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material 3-Amino-4-methylpyridine
compliance-icons compliance-icons
Product Identifier
  • 3-Amino-4-methylpyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.