missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylbenzoic Acid, TRC

Product Code. 30136929
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136929 100 g 100g
30134396 250 g 250g
17990995 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136929 Supplier Toronto Research Chemicals Supplier No. TRCA614090100G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2458-12-0
Molecular Formula C8 H9 N O2
Molecular Weight (g/mol) 151.16
Synonym Benzoic acid, 3-amino-4-methyl-, p-Toluic acid, 3-amino- (6CI,7CI,8CI), 3-Amino-4-methylbenzoic acid, 2-Toluidine-4-carboxylic acid, 4-Methyl-3-aminobenzoic acid, NSC 127033, NSC 597, 3-Amino-4-methylbenzoic Acid
IUPAC Name 3-amino-4-methylbenzoic acid
SMILES Cc1ccc(cc1N)C(=O)O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H9NO2/c1-5-2-3-6(8(10)11)4-7(5)9/h2-4H,9H2,1H3,(H,10,11)
Formula Weight 151.0633
Chemical Name or Material 3-Amino-4-methylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.