missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylbenzoic Acid, Mikromol™

Product Code. 17930886
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17930886 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17930886 Supplier Mikromol Supplier No. MM3374.170025
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Amino-4-methylbenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Amino-4-methylbenzoic Acid
CAS 2458-12-0
Analyte or Component Names 3-Amino-4-methylbenzoic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O2
Certifications/Compliance ISO 9001
InChI Formula InChI=1S/C8H9NO2/c1-5-2-3-6(8(10)11)4-7(5)9/h2-4H,9H2,1H3,(H,10,11)
SMILES Cc1ccc(cc1N)C(=O)O
IUPAC Name 3-amino-4-methylbenzoic acid
Molecular Weight (g/mol) 151.16
Quantity 25 mg
Formula Weight 151.0633 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.