missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylbenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 11388636
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388636 1 g 1g
11398636 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388636 Supplier Thermo Scientific Alfa Aesar Supplier No. L17695.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22237-12-3
Molecular Formula C7H11BClNO2
Molecular Weight (g/mol) 187.43
MDL Number MFCD01632201
InChI Key BKFDOPYYSBQWIK-UHFFFAOYSA-N
Synonym 3-amino-4-methylphenyl boronic acid, 3-amino-4-methylbenzeneboronic acid, 3-amino-4-methyl-phenyl boronic acid, 3-amino-p-tolylboronic acid, boronic acid, 3-amino-4-methylphenyl, acmc-20agxu, pubchem1745, 3-amino-4-methylphenylboranic acid, 3-amino4methyl phenyl boronic acid
PubChem CID 2737803
IUPAC Name (3-amino-4-methylphenyl)boronic acid
SMILES Cl.CC1=CC=C(C=C1N)B(O)O

Specifications

Melting Point ∼250°C
Quantity 1 g
Formula Weight 150.98
Percent Purity 98%
Chemical Name or Material 3-Amino-4-methylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.