missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylbenzamide, TRC

Product Code. 30137687
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137687 25 g 25g
30003297 2.5 g 2.5g
30011579 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137687 Supplier Toronto Research Chemicals Supplier No. TRCA61467325G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19406-86-1
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
Synonym Benzamide, 3-amino-4-methyl- (9CI, ACI), 3-Amino-4-methylbenzamide (ACI), p-Toluamide, 3-amino- (8CI), M 60
IUPAC Name 3-amino-4-methylbenzamide
SMILES Cc1ccc(cc1N)C(=O)N

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H10N2O/c1-5-2-3-6(8(10)11)4-7(5)9/h2-4H,9H2,1H3,(H2,10,11)
Formula Weight 150.0793
Chemical Name or Material 3-Amino-4-methylbenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.