missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-iodopyridine, 98%

Product Code. 13575030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13575030 1 g 1g
13585030 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13575030 Supplier Thermo Scientific Acros Supplier No. 448840010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 105752-11-2 | C5H5IN2 | 220.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105752-11-2
Molecular Formula C5H5IN2
Molecular Weight (g/mol) 220.01
MDL Number MFCD04971334
InChI Key ZJRSKTXMSIVNAU-UHFFFAOYSA-N
Synonym 3-amino-4-iodopyridine, 4-iodo-pyridin-3-ylamine, 3-pyridinamine, 4-iodo, 4-iodo-3-pyridinamine, 3-amine-4-iodopyridine, pubchem6681, 4-iodo-3-pyridylamine, 4-iodo-3-aminopyridine, 4-iodo-3-pyridinylamine, 4-iodopyridin-3-ylamine
PubChem CID 819132
IUPAC Name 4-iodopyridin-3-amine
SMILES NC1=CN=CC=C1I

Specifications

Melting Point 65°C to 70°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 3-Amino-4-iodopyridine
compliance-icons compliance-icons
Product Identifier
  • 3-Amino-4-iodopyridine
Signal Word
  • Danger||Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.