missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-chlorobenzoic acid, 98%, Thermo Scientific™

Product Code. 15750839 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15750839 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15750839 Supplier Thermo Scientific Acros Supplier No. 219305000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2840-28-0
Melting Point 214.0°C to 218.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C7H6ClNO2
Linear Formula H2NC6H3(Cl)CO2H
Quantity 500 g
Beilstein 14,412
Synonym benzoic acid, 3-amino-4-chloro, unii-3q8r4430cf, 3-amino-4-chloro benzoic acid, 3-amino-4-chloro-benzoic acid, 4-chloro-3-amino benzoic acid, pubchem8988, acmc-1cmgg, 5-carboxy-2-chloroaniline, dsstox_cid_24414, dsstox_rid_80213
InChI Key DMGFVJVLVZOSOE-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(=O)O)N)Cl
IUPAC Name 3-amino-4-chlorobenzoic acid
Molecular Weight (g/mol) 171.58
PubChem CID 76092
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 3-Amino-4-chlorobenzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.