missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-chlorobenzamide, 97%

Product Code. 11490878
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490878 5 g 5g
11400888 25 g 25g
11410888 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490878 Supplier Thermo Scientific Alfa Aesar Supplier No. B25294.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19694-10-1 | C7H7ClN2O | 170.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19694-10-1
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.60
MDL Number MFCD00035785
InChI Key QHMDKGRWJVOUFU-UHFFFAOYSA-N
PubChem CID 88200
IUPAC Name 3-amino-4-chlorobenzamide
SMILES NC(=O)C1=CC=C(Cl)C(N)=C1

Specifications

Melting Point 164°C to 166°C
Quantity 5 g
Synonym 3-amino-4-chloro-benzamide, benzamide, 3-amino-4-chloro, fast red base db-30, acmc-209f0q, 3-amino-4-chloro benzamide, benzamide,3-amino-4-chloro, 3-azanyl-4-chloranyl-benzamide, 3-amino-4-chlorobenzamide, 3-amino-4-chlorobenzoic acidamide
Formula Weight 170.6
Percent Purity 97%
Chemical Name or Material 3-Amino-4-chlorobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.