missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-bromo-1H-pyrazole, 97%, Thermo Scientific Chemicals

Product Code. 11464483
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16461-94-2
Molecular Formula C3H4BrN3
Molecular Weight (g/mol) 161.99
MDL Number MFCD00082728
InChI Key OELYMZVJDKSMOJ-UHFFFAOYSA-N
Synonym 3-amino-4-bromopyrazole, 4-bromo-1h-pyrazol-3-amine, 5-amino-4-bromo-1h-pyrazole, 3-amino-4-bromo-1h-pyrazole, 1h-pyrazol-3-amine, 4-bromo, 4-bromo-2h-pyrazol-3-amine, 4-bromo-1h-pyrazol-3-ylamine, 4-bromo-2h-pyrazol-3-ylamine, 4-bromopyrazole-5-ylamine, 3-amino-4-bromo pound inverted question markpyrazole
PubChem CID 140079
IUPAC Name 4-bromo-1H-pyrazol-5-amine
SMILES C1=NNC(=C1Br)N

Specifications

Melting Point 134°C to 137°C
Odor Odorless
Quantity 5 g
Beilstein 1617109
Formula Weight 161.99
Percent Purity 97%
Chemical Name or Material 3-Amino-4-bromo-1H-pyrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'