missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-bromo-1H-indazole, 95%, Thermo Scientific™

Product Code. 10189969 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10189969 1 g 1g
10833391 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10189969 Supplier Thermo Scientific Acros Supplier No. 435790010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 914311-50-5
Molecular Formula C7H6BrN3
Molecular Weight (g/mol) 212.05
MDL Number MFCD09909580
InChI Key IXXXOIXGQFPLIH-UHFFFAOYSA-N
Synonym 3-amino-4-bromo-1h-indazole, 4-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 4-bromo, 3-amino-4-bromoindazole, pubchem20681, acmc-209rbi, 4-bromo-1h-indazole-3-amine, 4-bromo-2h-indazol-3-ylamine, 4-bromo-1h-indazole-3-ylamine
PubChem CID 11975123
IUPAC Name 4-bromo-1H-indazol-3-amine
SMILES NC1=NNC2=CC=CC(Br)=C12

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 147°C to 152°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 95%
Quantity 1 g
Formula Weight 212.05
Percent Purity ≥94.0%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-bromo-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-4-bromo-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.