missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-oxo-propanoic Acid Phenylmethyl Ester, TRC

Product Code. 30310769
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310769 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310769 Supplier Toronto Research Chemicals Supplier No. TRCA61222550MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Amino-3-oxo-propanoic Acid Phenylmethyl Ester
CAS 1042348-44-6
Molecular Formula C11 H15 N5 O5
Quantity 50 mg
InChI Formula InChI=1S/C11H15N5O5/c1-11(20)6(18)4(2-17)21-9(11)16-3-13-5-7(16)14-10(12)15-8(5)19/h3-4,6,9,17-18,20H,2H2,1H3,(H3,12,14,15,19)/t4-,6?,9-,11?/m1/s1
SMILES CC1(O)C(O)[C@@H](CO)O[C@H]1n2cnc3C(=O)N=C(N)Nc23
IUPAC Name 2-amino-9-[(2R,5R)-3,4-dihydroxy-5-(hydroxymethyl)-3-methyloxolan-2-yl]-3H-purin-6-one
Molecular Weight (g/mol) 297.267
Formula Weight 297.107
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.