missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-(2-chlorophenyl)propionic acid, 97%, Thermo Scientific™

Product Code. 11399755
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399755 1 g 1g
11309765 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399755 Supplier Thermo Scientific Alfa Aesar Supplier No. L19796.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68208-20-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD01871299
InChI Key NXXFYRJVRISCCP-MRVPVSSYSA-N
Synonym 3-amino-3-2-chlorophenyl propanoic acid, 3-amino-3-2-chloro-phenyl-propionic acid, 3-2-chlorophenyl-dl-beta-alanine, 3-amino-3-2-chlorophenyl propionic acid, dl-beta-2-chlorophenyl alanine, pubchem13916, rarechem ak hc t319, 3-amino-3-2-chlorophenyl propanoicacid, benzenepropanoic acid, .beta.-amino-2-chloro, r/s-3-amino-3-2-chloro-phenyl-propionic acid
PubChem CID 239929
IUPAC Name 3-amino-3-(2-chlorophenyl)propanoic acid
SMILES [NH3+][C@H](CC([O-])=O)C1=CC=CC=C1Cl

Specifications

Melting Point ∽223°C (decomposition)
Beilstein 2938797
Quantity 1 g
Formula Weight 199.64
Percent Purity 97%
Chemical Name or Material 3-Amino-3-(2-chlorophenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.